Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

D-(-)-Sorbitol

Glass bottle for solids, 1 kg
Catalog # MOLEM26965258
Marketsource
Supplier:   Molekula
D-(-)-Sorbitol
Catalog # MOLEM26965258
Supplier:  Molekula
Marketsource
CAS Number:  50-70-4
D-(-)-Sorbitol
D-(-)-Sorbitol

Glass bottle for solids, 1 kg

CAS Number:  50-70-4
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,08797
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:94…98 °C
  • Density:1.524 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:200-061-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.