Order Entry
Belgium
ContactUsLinkComponent
D-(-)-Mannitol 97.0-102.0% Ph. Eur., USP
D-(-)-Mannitol 97.0-102.0% Ph. Eur., USP
Supplier:  VWR Chemicals
Certificates
undefined
D-(-)-Mannitol 97.0-102.0% Ph. Eur., USP
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    Conforms (see CoA/CoS)
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Residual solvents:
    Unlikely by manuf.process
  • Metal catalysts:
    Unlikely by manuf.process
  • As (Arsenic):
    Max. 2 ppm
  • Cd (Cadmium):
    Max. 1 ppm
  • Co (Cobalt):
    Max. 1 ppm
  • Hg (Mercury):
    Max. 2 ppm
  • Pb (Lead):
    Max. 3 ppm
  • V (Vanadium):
    Max. 1 ppm
  • Conforms to Ph.Eur./USP:
    Passes test
  • Ph.Eur. specifications::
    Max. 2.0 %
  • Assay (dried substance) [Ph.Eur.]:
    97.0 --> 102.0 %
  • Identification C:
    Passes test Ph.Eur.
  • Appearance of solution:
    Passes test Ph.Eur.
  • Conductivity (20°C; 20 %) [Ph.Eur.]:
    Max. 20 μS/cm
  • Melting point [Ph.Eur.]:
    165 --> 170 °C
  • Reducing sugars (as C6H12O6) [Ph.Eur.]:
    Max. 0.1 %
  • D-Sorbitol [Ph.Eur.]:
    Max. 2.0 %
  • D-Maltitol and isomalt [Ph.Eur.]:
    Max. 2.0 %
  • Unspecified impur. (for each) [Ph.Eur.]:
    Max. 0.10 %
  • Total impurities [Ph.Eur.]:
    Max. 2.0 %
  • Disregard limit [Ph.Eur.]:
    Max. 0.05 %
  • Loss on drying (105°C; 4 h) [Ph.Eur.]:
    Max. 0.5 %
  • TAMC [Ph.Eur.]:
    Max. 1000 CFU/g
  • TYMC [Ph.Eur.]:
    Max. 100 CFU/g
  • Escherichia coli [Ph.Eur.]:
    Absent
  • Salmonella [Ph.Eur.]:
    Absent
  • Assay (calculated on dried basis) [USP]:
    97.0 --> 102.0 %
  • Identification A:
    Passes test USP
  • Disregard limit [USP]:
    Max. 0.05 %
  • Sorbitol [USP]:
    Max. 2.0 %
  • Sum of isomalt and maltitol [USP]:
    Max. 2.0 %
  • Unspecified impurities [USP]:
    Max. 0.10 %
  • Total impurities [USP]:
    Max. 2.0 %
  • Reducing sugars (as C6H12O6) [USP]:
    Max. 0.1 %
  • Ni (Nickel) [USP]:
    Max. 1 μg/g
  • Melting point [USP]:
    165 --> 170 °C
  • Loss on drying (105°C; 4 h) [USP]:
    Max. 0.5 %
  • Conductivity (25°C; 20 %) [USP]:
    Max. 20 μS/cm
  • TAMC [USP]:
    Max. 1000 CFU/g
  • TYMC [USP]:
    Max. 100 CFU/g
  • Escherichia coli [USP]:
    Absent

Specification Test Results

Frequently Bought Together