Order Entry
Belgium
ContactUsLinkComponent
L(+)-Potassium sodium tartrate tetrahydrate, TECHNICAL
L(+)-Potassium sodium tartrate tetrahydrate, TECHNICAL
Catalog # 27062.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
L(+)-Potassium sodium tartrate tetrahydrate, TECHNICAL
Catalog # 27062.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
 :282.22 g/mol
Merck Index:13,07755
Melting Pt:70…80 °C
Density:∼1.767 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-156-8
REACH:01-2119524666-34
CAS Number:6381-59-5
Formula:KOOC(CHOH)₂COONa·4H₂O
MDL Number:MFCD00150989