L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0% ACS
:6381-59-5
:
:KOOC(CHOH)₂COONa·4H₂O
:MFCD00150989
:282.22 g/mol
:14,7670
:70…80 °C
:1.77
:Ambient
:206-156-8
- Assay (unspecified):99.0-102.0% C₄H₄KNaO₆.4H₂O
- Impurity content:Ca 0.005%
- Insoluble material:0.005%
- pH of a 5% solution:6.0-8.5 at 25°C
- Trace Metals:(as Pb) 5ppm
Frequently Bought Together