Order Entry
Belgium
Orders LinkContactUsLinkComponent
L(-)-Malic acid

L(-)-Malic acid

Irritant
Marketsource
Supplier:   Serva
L(-)-Malic acid
Supplier:  Serva
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
  • CAS Number:97-67-6
  • Formula:C₄H₆O₅
  • MDL Number:MFCD00064213
  • Molecular Weight:134.09 g/mol
  • Merck Index:13,05730
  • Boiling Pt:322 °C (1013 hPa)
  • Melting Pt:101…104 °C
  • Density:1.595 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:202-601-5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

    Specifications:
    Marketsource
    Marketsource