Order Entry
Belgium
Orders LinkContactUsLinkComponent
L(-)-Malic acid

L(-)-Malic acid

Irritant
Marketsource
Supplier:   Roth Carl
L(-)-Malic acid
Supplier:  Roth Carl
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
  • CAS Number:97-67-6
  • Formula:C₄H₆O₅
  • MDL Number:MFCD00064213
  • Molecular Weight:134.09 g/mol
  • Merck Index:13,05730
  • Boiling Pt:322 °C (1013 hPa)
  • Melting Pt:101…104 °C
  • Density:1.595 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:202-601-5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.