Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(+)-Malic acid
D-(+)-Malic acid
D-(+)-Malic acid
MOLE48772307-5G
:
Molekula
:
D-(+)-Malic acid
MOLE48772307-5G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
134.09 g/mol
:
13,05307
:
322 °C (1013 hPa)
:
101…104 °C
:
Ambient
:
211-262-2
:
GHS07
:
636-61-3
:
C₄H₆O₅
:
MFCD00004245