Order Entry
Belgium
ContactUsLinkComponent
D-(+)-Malic acid for synthesis, Sigma-Aldrich®
D-(+)-Malic acid for synthesis, Sigma-Aldrich®
Catalog # 8.00763.0005
Supplier:  Merck
CAS Number:  
D-(+)-Malic acid for synthesis, Sigma-Aldrich®
Catalog # 8.00763.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Solubility:
    364 g/l (20 °C)
  • Melting Point:
    101 - 104 °C
  • Molar Mass:
    134.08 g/mol
  • Bulk Density:
    600 kg/m3
  • pH:
    2.2 (10 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :134.09 g/mol
Merck Index:13,05307
Boiling Pt:322 °C (1013 hPa)
Melting Pt:101…104 °C
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:211-262-2
 :GHS07
CAS Number:636-61-3
Formula:C₄H₆O₅
MDL Number:MFCD00004245