Organic reference standard, 2,4,4',5-Tetrachlorobiphenyl (PCB No. 74) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20007400IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:32690-93-0
- Description:2,4,4',5-Tetrachlorobiphenyl (PCB No. 74) 10 µg/ml in isooctane
- Environmentally Preferable:
- Formula:C₁₂H₆Cl₄
- Description:2,4,4',5-Tetrachlorobiphenyl (PCB No. 74)