Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, 2,2',3,5,6,6'-Hexachlorobiphenyl (PCB No. 152) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3,5,6,6'-Hexachlorobiphenyl (PCB No. 152) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-152
CAS Number:  
Organic reference standard, 2,2',3,5,6,6'-Hexachlorobiphenyl (PCB No. 152) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-152
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    68194-09-2
  • Description:
    2,2',3,5,6,6'-Hexachlorobiphenyl (PCB No. 152) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₄Cl₆
  • Description:
    2,2',3,5,6,6'-Hexachlorobiphenyl (PCB No. 152)

Specifications

About this item