Order Entry
Belgium
ContactUsLinkComponent
(S)-(+)-Arginine hydrochloride ≥99% for synthesis, Sigma-Aldrich®
(S)-(+)-Arginine hydrochloride ≥99% for synthesis, Sigma-Aldrich®
Catalog # 8.16001.0100
Supplier:  Merck
CAS Number:  
(S)-(+)-Arginine hydrochloride ≥99% for synthesis, Sigma-Aldrich®
Catalog # 8.16001.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay (argentometric):
    ≥ 99.0 %
  • Spec. rotation [α²0/D (c=8 in HCl 6 mol/l):
    +21.5 - +23.0 °
  • Identity (IR):
    passes test
  • Bulk Density:
    1250 kg/m3
  • Density:
    1.42 g/cm3 (20 °C)
  • pH:
    5.7 (100 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :210.66 g/mol
Merck Index:13,00785
Melting Pt:218 °C
Density:1.42 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:214-275-1
CAS Number:1119-34-2
Formula:C₆H₁₅ClN₄O₂
MDL Number:MFCD00064550