Order Entry
Belgium
ContactUsLinkComponent
L-Arginine hydrochloride 98.0-102.0% (by titrimetric analysis on dry substance), BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
L-Arginine hydrochloride 98.0-102.0% (by titrimetric analysis on dry substance), BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
L-Arginine hydrochloride 98.0-102.0% (by titrimetric analysis on dry substance), BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
Supplier:  Avantor

Specification Test Results

  • Assay (C₆H₁₄N₄O₂HCl) (dried basis, by non-aqueous acid- base titration):
    98.0 - 102.0 %
  • Specific Rotation,α²ºD(dried basis, c=80mg in 6N HCL)(+):
    21.4 - 23.6 Degree
  • Homogeneity by TLC:
    Passes Test
  • Ash (sulfated):
    ≤ 0.1 %
  • Loss on Drying at 105°C:
    ≤ 0.5 %
  • Heavy Metals (as Pb):
    ≤ 0.002 %
  • Iron (Fe):
    ≤ 0.005 %

Specification Test Results

Frequently Bought Together