Organic reference standard, 2,3',4,4',5',6-Hexachlorobiphenyl (PCB No. 168) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20016800IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:59291-65-5
- Description:2,3',4,4',5',6-Hexachlorobiphenyl (PCB No. 168) 10 µg/ml in isooctane
- Environmentally Preferable:
- Formula:C₁₂H₄Cl₆
- Description:2,3',4,4',5',6-Hexachlorobiphenyl (PCB No. 168