Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, 3,3',4,5'-Tetrachlorobiphenyl (PCB No. 79) 100 µg/ml in isooctane, CIL
Organic reference standard, 3,3',4,5'-Tetrachlorobiphenyl (PCB No. 79) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-79-CS
CAS Number:  
Organic reference standard, 3,3',4,5'-Tetrachlorobiphenyl (PCB No. 79) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-79-CS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    41464-48-6
  • Description:
    3,3',4,5'-Tetrachlorobiphenyl (PCB No. 79) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₆Cl₄
  • Description:
    3,3',4,5'-Tetrachlorobiphenyl (PCB No. 79)

Specifications

About this item