
L(+)-Arginine pour biochimie, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:74-79-3
:
Formula:C₆H₁₄N₄O₂
MDL Number:MFCD00002635
:174.2 g/mol
Merck Index:13,00785
Boiling Pt:368 °C (1013 hPa)
Melting Pt:238 °C
Density:1,3 g/cm³ (20 °C)
:M-Clarity™ (Niveau de qualité) = MQ100
EINECS:200-811-1
:GHS07
Storage Temperature:Température ambiante
Related Information
Certificates
Search by Lot Numbermsds_101542
SDS
ProductPage_101542 ProductPage_101542
Product specification
Frequently Bought Together