Organic reference standard, Arginine Aspartate, Ph. Eur.
Catalog # EDQMY0000304
Supplier: EDQM
CAS Number:
Specifications
- Pk:20 mg
- CAS No.:7675-83-4
- Description:Arginine aspartate, Ph. Eur. standard
- Formula:C₁₀H₂₁N₅O₆
- Description:Arginine aspartate, Ph. Eur. standard
Specifications
About this item
Storage conditions: Keep refrigerated.