Yellow IHC chromogen (HRP), HighDef®
Catalog # ENZOADI9501700030
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Cdt:30 mL
- Désignation:Yellow IHC chromogen (HRP)
- Seq:02
- Conditions de stockage:Long term
- Température de stockage:4 °C
Specifications
About this item
Yellow IHC chromogen (HRP) is intended to be used in conjunction with horseradish peroxidase (HRP)-based immunostaining or in situ hybridisation systems.
- High definition yellow IHC chromogen for HRP-activated development
- Distinct bright yellow that can easily be distinguished from other stains
- This system can be used in any automation system as well as manual use
Delivery: Substrate buffer and chromogen in an empty mixing bottle.