Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

L(+)-Glutamine, TECHNICAL

Plastic bottle for solids, 100 g
Catalog # 24378.187
Supplier:   VWR Chemicals
L(+)-Glutamine, TECHNICAL
Catalog # 24378.187
Supplier:  VWR Chemicals
CAS Number:  56-85-9
L(+)-Glutamine, TECHNICAL
L(+)-Glutamine, TECHNICAL

Plastic bottle for solids, 100 g

CAS Number:  56-85-9
Price
85,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
  • CAS Number:56-85-9
  • Formula:C₅H₁₀N₂O₃
  • MDL Number:MFCD00008044
  • Molecular Weight:146.15 g/mol
  • Merck Index:13,04484
  • Boiling Pt:355 °C (1013 hPa)
  • Melting Pt:185…186 °C
  • Density:1.5326 g/cm³ (130 °C)
  • Storage Temperature:Ambient
  • EINECS:200-292-1
Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Cat. No.:
    24378.187

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.