L(+)-Glutamine, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:56-85-9
:
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00008044
:146.15 g/mol
Merck Index:13,04484
Boiling Pt:355 °C (1013 hPa)
Melting Pt:185…186 °C
Density:1.5326 g/cm³ (130 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:200-292-1
Related Information
Frequently Bought Together