Organic reference standard, cis-Dichloroethylene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12422500ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:156-59-2
- Description:cis-Dichloroethylene 100 µg/ml in methanol
- Formula:C₂H₂Cl₂
- Description:cis-1,2-Dichloroethene