(R)-(-)-Linalool, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: (-)-Linalool, (R)-3,7-Dimethylocta-1,6-dien-3-ol, (R)-(-)-3,7-Dimethyl-1,6-octadien-3-ol, L-Linalool
CAS Number:126-91-0
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00135469
Molecular Weight:154.25 g/mol
:M-Clarity™ quality level = MQ100
EINECS:204-811-2
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together