Pesticide reference standard, 1,2,3-Trichloropropane 2000 µg/ml in methanol
Supplier: Restek
Certificates
About this item
Specifications
- CAS No.:96-18-4
- Formula:C₃H₅Cl₃
- Description:1,2,3-Trichloropropane
Specifications
Frequently Bought Together