Order Entry
Belgium
ContactUsLinkComponent
Penicillin : Streptomycin solution 0.06/0.1 g/L
Penicillin : Streptomycin solution 0.06/0.1 g/L
Catalog # L0018-100
Supplier:  Biowest
CAS Number:  
undefined
Penicillin : Streptomycin solution 0.06/0.1 g/L
Catalog # L0018-100
Supplier:  Biowest
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
Storage Temperature:Freezer
CAS Number:1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)
 :113-98-4