Order Entry
Belgium
ContactUsLinkComponent
Iron(II) sulphate hydrate ≥84%, GPR RECTAPUR®
Irritant
Iron(II) sulphate hydrate ≥84%, GPR RECTAPUR®
Catalog # 24242.292
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Iron(II) sulphate hydrate ≥84%, GPR RECTAPUR®
Catalog # 24242.292
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 84 %
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Insolubility in water:
    Max. 0.05 %
  • As (Arsenic):
    Max. 2 ppm
  • Fe (III) (Iron):
    Max. 0.4 %
  • Pb (Lead):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :169.93 g/mol
Melting Pt:∼300 °C
Density:2.97 g/cm³ (20 °C)
EINECS:231-753-5
 :GHS07
Storage Temperature:Ambient
CAS Number:13463-43-9
Formula:FeSO₄·H₂O
MDL Number:MFCD00149722