Order Entry
Belgium
ContactUsLinkComponent
IHC universal negative control reagent
IHC universal negative control reagent
Catalog # ENZOADI9502310025
Supplier:  Enzo Life Sciences
CAS Number:  
IHC universal negative control reagent
Catalog # ENZOADI9502310025
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Description:
    IHC universal negative control reagent
  • Seq:
    02
  • Form:
    Liquid
  • Storage temperature:
    4 °C
  • Application:
    IHC

Specifications

About this item

IHC universal negative control reagent is specifically designed to work with rabbit, mouse and goat antibodies.

  • Liquid mixture of purified goat, mouse and rabbit immunoglobulins (IgGs)

The reagent has been titered to work with polymer-based secondary systems in manual and automated IHC platforms.