Order Entry
Belgium
ContactUsLinkComponent
IHC chromogen substrate (AEC, stable), HIGHDEF®
IHC chromogen substrate (AEC, stable), HIGHDEF®
Catalog # ENZOADI9502100030
Supplier:  Enzo Life Sciences
CAS Number:  
IHC chromogen substrate (AEC, stable), HIGHDEF®
Catalog # ENZOADI9502100030
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    30 mL
  • Description:
    IHC chromogen liquid substrate (AEC, stable)
  • Seq:
    02
  • Storage conditions:
    Protect from light
  • Form:
    Liquid substrate (AEC, stable)
  • Storage temperature:
    4 °C
  • Colour:
    Amber

Specifications

About this item

HIGHDEF® red IHC chromogen can be used in conjunction with peroxidase-based immunostaining systems.

Peroxidase from the immunodetection system reacts with hydrogen peroxide substrate to degrade it, which reacts with AEC precipitating it at positive sites yielding a reddish brown product.