Order Entry
Belgium
ContactUsLinkComponent
VWR®, DNA marker, 1 kb
VWR®, DNA marker, 1 kb
Catalog # K181-1ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, DNA marker, 1 kb
Catalog # K181-1ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Description:
    DNA marker, 1 kb ladder

Specifications

About this item

DNA marker consists of 11 fragments ranging from 500 to 10000 bp for easy band identification.

  • Sufficient for 150 lanes
  • Available pre-mixed with loading dye or with EZ-Vision® One DNA dye