Order Entry
Belgium
ContactUsLinkComponent
Iron(III) nitrate nonahydrate
Oxidizing
Irritant
Iron(III) nitrate nonahydrate
Catalog # LOHM7543600.9025
Supplier:  LOHMANN
CAS Number:  
Oxidizing
Irritant
Iron(III) nitrate nonahydrate
Catalog # LOHM7543600.9025
Supplier:  LOHMANN
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :404 g/mol
Merck Index:13,04057
Boiling Pt:125 °C (1013 hPa)
Melting Pt:47 °C
Density:1.68 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-899-5
UN:1466
ADR:5.1,III
 :GHS03
 :GHS07
CAS Number:7782-61-8
Formula:Fe(NO₃)₃·9H₂O
MDL Number:MFCD00149708