ContactUsLinkComponent
IHC tissue primer
IHC tissue primer
Catalog # ENZOADI9502340100
Supplier:  Enzo Life Sciences
CAS Number:  
IHC tissue primer
Catalog # ENZOADI9502340100
Supplier:  Enzo Life Sciences
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    IHC tissue primer
  • Seq:
    02
Specifications
About this item
Immunohistochemistry tissue primer is an enzyme blocking reagent.
  • TRIS-based alkaline solution with 30% H₂O₂
Designed for use in horse radish peroxidase (HRP) and alkaline phosphatase (AP) based immunohistochemical staining procedures on paraffin tissue sections, frozen tissue sections and cell smears to suppress non specific staining due to endogenous enzyme activity.