Manganese(II) sulphate monohydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
: Manganese sulphate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate
:10034-96-5
:
:MnSO₄·1H₂O
:MFCD00149159
:13,05763
:700 °C
:2.95 g/cm³ (25 °C)
:Ambient
:9,III
:M-Clarity™ quality level = MQ300
:169.02 g/mol
:232-089-9
:3077
:GHS05
:GHS08
:GHS09
- Environmentally Preferable:
- Solubility:762 g/l (20 °C)
- Melting Point:700 °C (anhydrous substance)
- Molar Mass:169.02 g/mol
- Bulk Density:1000 - 1200 kg/m3
- Density:2.95 g/cm3 (20 °C)
- pH:3.0 - 3.5 (50 g/l, H2O, 20 °C)
- Hazard Class:9
- Hazard Class Description:Miscellaneous hazardous material
Frequently Bought Together