

Manganese(II) chloride tetrahydrate 99.0-101.0%, AnalaR NORMAPUR® ACS analytical reagent
:13446-34-9
:
:MnCl₂·4H₂O
:MFCD00149792
:197.9 g/mol
:13,05751
:1190 °C (1013 hPa)
:58…59 °C
:2.01 g/cm³ (20 °C)
:Ambient
:231-869-6
:3077
:9,III
:GHS07
:GHS09
- Assay:99.0 - 101.0 %
- pH (25°C; 5 %):3.5 - 6.0
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 50 ppm
- Not precipitated by (NH4)2S (as SO4):Max. 0.4 %
- Reducing substances (as O):Max. 5 ppm
- SO4 (Sulphate):Max. 50 ppm
- Ca (Calcium):Max. 50 ppm
- Co (Cobalt):Max. 20 ppm
- Cu (Copper):Max. 5 ppm
- K (Potassium):Max. 100 ppm
- Fe (Iron):Max. 5 ppm
- Mg (Magnesium):Max. 50 ppm
- Na (Sodium):Max. 0.05 %
- Pb (Lead):Max. 5 ppm
- Zn (Zinc):Max. 50 ppm
- Conforms to ACS:Passes test
Frequently Bought Together