Order Entry
Belgium
ContactUsLinkComponent
Manganese(II) chloride tetrahydrate, EMSURE® ACS for analysis, Supelco®
Toxic
Corrosive
Health
Environment
Manganese(II) chloride tetrahydrate, EMSURE® ACS for analysis, Supelco®
Supplier:  Merck
Certificates
Toxic
Corrosive
Health
Environment
Manganese(II) chloride tetrahydrate, EMSURE® ACS for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (complexometric):
    99.0 - 101.0 %
  • Insoluble matter:
    ≤ 0.005 %
  • pH-value (5 %; water; 25 °C):
    3.5 - 6.0
  • Sulfate (SO₄):
    ≤ 0.002 %
  • Heavy metals (as Pb):
    ≤ 0.0005 %
  • Ca (Calcium):
    ≤ 0.001 %
  • Cu (Copper):
    ≤ 0.0005 %
  • Fe (Iron):
    ≤ 0.0005 %
  • K (Potassium):
    ≤ 0.001 %
  • Mg (Magnesium):
    ≤ 0.005 %
  • Na (Sodium):
    ≤ 0.005 %
  • Pb (Lead):
    ≤ 0.0005 %
  • Zn (Zinc):
    ≤ 0.001 %
  • Substances not precipitated by ammonium sulfide (as SO₄):
    ≤ 0.2 %
  • Substances reducing potassium permanganate (as O):
    ≤ 0.0005 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together