Mercury(II) nitrate monohydrate ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Mercury(II) nitrate hydrate, Mercury dinitrate monohydrate
CAS Number:7783-34-8
:13465-31-1
Formula:Hg(NO₃)₂·1H₂O
MDL Number:MFCD00149736
Density:4.3 g/cm³ (20 °C)
Molecular Weight:342.62 g/mol
Melting Pt:79
ADR:6.1 II
:M-Clarity™ quality level = MQ300
EINECS:233-152-3
UN:1625
:GHS06
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (complexometric):≥ 99.0 %
- Chloride (Cl):≤ 0.002 %
- Sulfate (SO₄):≤ 0.002 %
- Cu (Copper):≤ 0.0005 %
- Fe (Iron):≤ 0.001 %
- K (Potassium):≤ 0.005 %
- Na (Sodium):≤ 0.005 %
- Residue after reduction:≤ 0.01 %
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_104439
SDS
ProductPage_104439 ProductPage_104439
Product specification
Frequently Bought Together