Order Entry
Belgium
ContactUsLinkComponent
DNA isolation kit, EPIXTRACT®
DNA isolation kit, EPIXTRACT®
Supplier:  ENZO LIFE SCIENCES
Certificates
DNA isolation kit, EPIXTRACT®
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Biofluids (serum, plasma, saliva)
  • Target molecule:
    DNA
  • Description:
    EPIXTRACT® DNA isolation kit

Specifications

Frequently Bought Together