Order Entry
Belgium
ContactUsLinkComponent
Copper(II) sulphate, anhydrous ≥97%, TECHNICAL
Copper(II) sulphate, anhydrous ≥97%, TECHNICAL
Catalog # 23168.265
Supplier:  VWR Chemicals
CAS Number:  
undefined
Copper(II) sulphate, anhydrous ≥97%, TECHNICAL
Catalog # 23168.265
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 97 %

Specification Test Results

About this item

Synonyms: Cupric sulphate
 :
 :159.61 g/mol
Merck Index:12,02722
Melting Pt:200 °C
Density:3.603 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-847-6
UN:3288
ADR:6.1,III
 :GHS07
 :GHS09
CAS Number:7758-98-7
Formula:CuSO₄
MDL Number:MFCD00010981