Order Entry
Belgium
ContactUsLinkComponent
Mouse-HRP/rabbit-AP IHC, MULTIVIEW®
Mouse-HRP/rabbit-AP IHC, MULTIVIEW®
Catalog # ENZOADI9501000001
Supplier:  Enzo Life Sciences
CAS Number:  
Mouse-HRP/rabbit-AP IHC, MULTIVIEW®
Catalog # ENZOADI9501000001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Mouse-HRP/rabbit-AP IHC
  • Seq:
    02
  • Storage conditions:
    Long term
  • Storage temperature:
    4 °C

Specifications

About this item

Mouse-HRP/rabbit-AP IHC is a biotin-free one-step detection system suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and cryostat sections.

  • Nanopolymer-based system for superior multiplex antigen detection
  • Expandable with additional chromogens
  • Preserves precious tissue while reducing reagents

May be used with blood smears, cytosmears, and cell preparations.

Ordering information: Includes a peroxidase block, POLYVIEW® IHC reagent (mouse-HRP), POLYVIEW® IHC reagent (rabbit-AP), HIGHDEF® IHC chromogen substrate (DAB, stable) buffer and chromogen, HIGHDEF® red IHC chromogen (AP) buffer and chromogen, empty mixing bottles.