Orders LinkContactUsLinkComponent
Low temperature covers
Supplier:   OHAUS
Low temperature covers
Supplier:  OHAUS
Certificates
Low temperature covers
Low temperature covers
3 Options Available Below
Product Details & Documents
Plexiglass cover reduces air flow for additional temperature stability in low temperature applications.
  • Two sides are open 6,4 mm
Product Family Options
Product Information
  • For
    W×D×H
    Availability
    Price
  • OHAU30400179
    Low temperature cover, plexiglass
    Low temperature cover, plexiglass
    2 blocks
    165×165×41 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 131,00 €
    131,00 €
    1 * 1 ST
       131,00 €
  • OHAU30400180
    Low temperature cover, plexiglass
    Low temperature cover, plexiglass
    4 blocks
    216×216×41 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 127,00 €
    127,00 €
    1 * 1 ST
       127,00 €
  • OHAU30400181
    Low temperature cover, plexiglass
    Low temperature cover, plexiglass
    6 blocks
    318×216×41 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 134,00 €
    134,00 €
    1 * 1 ST
       134,00 €