Orders LinkContactUsLinkComponent
Sandbaths for modular heating blocks
Sandbaths for modular heating blocks
Supplier:   OHAUS
Sandbaths for modular heating blocks
Supplier:  OHAUS
Certificates
Sandbaths for modular heating blocks
Sandbaths for modular heating blocks
4 Options Available Below
Product Details & Documents
Ideal for irregular shaped vessels.
  • Stainless steel construction for superior corrosion resistance
  • Designed to hold sand, stainless steel shot or non-volatile liquids
Product Family Options
Product Information
  • Availability
    Price
  • OHAU30400173
    Sand bath for one-block heater, 76×95×64 mm
    Sand bath for one-block heater, 76×95×64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 323,00 €
    323,00 €
    1 * 1 ST
       323,00 €
  • OHAU30400174
    Sand bath for two-block heater, 149×95×64 mm
    Sand bath for two-block heater, 149×95×64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 335,00 €
    335,00 €
    1 * 1 ST
       335,00 €
  • OHAU30400175
    Sand bath for four-block heater, 191×147×64 mm
    Sand bath for four-block heater, 191×147×64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 355,00 €
    355,00 €
    1 * 1 ST
       355,00 €
  • OHAU30400176
    Sand bath for six-block heater, 287×147×64 mm
    Sand bath for six-block heater, 287×147×64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 369,00 €
    369,00 €
    1 * 1 ST
       369,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Sand, 0,5 kg bag, for sand bath
    Sand, 0,5 kg bag, for sand bath
    OHAU30400177
    Log in for availability
    1 * 1 ST - 89,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Stainless steel shot, 0,5 kg, for sand bath
    Stainless steel shot, 0,5 kg, for sand bath
    OHAU30400178
    Log in for availability
    1 * 1 ST - 110,00 €