Ammonium iron(II) sulphate hexahydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate
CAS Number:7783-85-9
:
Formula:(NH₄)₂Fe(SO₄)₂·6H₂O
MDL Number:MFCD00150530
:392.14 g/mol
Melting Pt:110 °C
Density:1.864 g/cm³ (20 °C)
EINECS:233-151-8
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 99.0 %
- pH (25°C; 5 %):3.0 - 5.0
- Total P (Phosphorus):Max. 30 ppm
- Cl (Chloride):Max. 10 ppm
- Ca (Calcium):Max. 10 ppm
- Cu (Copper):Max. 10 ppm
- Fe (III) (Iron):Max. 0.03 %
- K (Potassium):Max. 50 ppm
- Mg (Magnesium):Max. 50 ppm
- Mn (Manganese):Max. 0,1 %
- Na (Sodium):Max. 50 ppm
- Pb (Lead):Max. 10 ppm
- Zn (Zinc):Max. 50 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250) 30L Plastic bucket UN Z (ZPACK13247037) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC24257.236 spec_VWRC24257.236 spec_VWRC24257.260 spec_VWRC24257.260 spec_VWRC24257.293
Product specification
More
Frequently Bought Together