Order Entry
Belgium
ContactUsLinkComponent
Ammonium heptamolybdate tetrahydrate ≥98.5%, GPR RECTAPUR®
Irritant
Ammonium heptamolybdate tetrahydrate ≥98.5%, GPR RECTAPUR®
Catalog # 21275.262
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Ammonium heptamolybdate tetrahydrate ≥98.5%, GPR RECTAPUR®
Catalog # 21275.262
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 98.5 %
  • Heavy metals (as Pb):
    Max. 50 ppm
  • PO4 + AsO4 + SiO4 (as PO4):
    Max. 0.02 %
  • Fe (Iron):
    Max. 50 ppm

Specification Test Results

About this item

 :
 :1235.86 g/mol
Merck Index:13,00536
Density:2.498 g/cm³ (20 °C)
EINECS:234-722-4
 :GHS07
Storage Temperature:Ambient
CAS Number:12054-85-2
 :12027-67-7
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00167059