Ammonium heptamolybdate tetrahydrate, Supelco®
: Merck
:12054-85-2
:12027-67-7
:(NH₄)₆Mo₇O₂₄·4H₂O
:MFCD00167059
:1235.86 g/mol
:13,00536
:2.498 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:234-722-4
:GHS07
:Ambient
- Solubility:400 g/l (20 °C)
- Melting Point:90 °C Elimination of water of crystallisation
- Molar Mass:1235.86 g/mol
- Bulk Density:800 kg/m3
- Density:2.498 g/cm3 (20 °C)
- pH:5.3 (50 g/l, H2O, 20 °C)
Frequently Bought Together