Order Entry
Belgium
ContactUsLinkComponent
Magnesium sulphate heptahydrate, SAFC®
Magnesium sulphate heptahydrate, SAFC®
Catalog # 1.72572.9025
CAS Number:  
Magnesium sulphate heptahydrate, SAFC®
Catalog # 1.72572.9025
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :246.48 g/mol
Merck Index:13,05715
Melting Pt:1124 °C
Density:2.66 g/cm³ (20 °C)
CAS Number:10034-99-8
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785