Order Entry
Belgium
ContactUsLinkComponent
Magnesium sulphate heptahydrate 99.0-100.5% (by complexometric analysis, on dry substance), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Magnesium sulphate heptahydrate 99.0-100.5% (by complexometric analysis, on dry substance), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Catalog # 1.05882.9500
CAS Number:  
Magnesium sulphate heptahydrate 99.0-100.5% (by complexometric analysis, on dry substance), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Catalog # 1.05882.9500
CAS Number:  

Specifications

  • Pk:
    500 kg
  • Seq:
    4826

Specifications

Specification Test Results

  • Assay (complexometric; calculated on dried substance):
    99.0 - 100.5 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • pH-value (5?%; water):
    5.0 - 9.2
  • Acidity or alkalinity:
    conforms
  • Chloride (Cl):
    ≤ 0.014%
  • As (Arsenic):
    ≤ 0.0002 %
  • Fe (Iron):
    ≤ 0.001 %
  • Se (Selenium):
    ≤ 0.001 %
  • Loss on drying (450?°C):
    49.0 - 52.0 %
  • Residual solvents (ICH Q3C):
    Excluded by production process

Specification Test Results

About this item

 :
 :246.48 g/mol
Merck Index:13,05715
Melting Pt:1124 °C
Density:2.66 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-298-2
CAS Number:10034-99-8
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785