Magnesium sulphate heptahydrate 99.0-100.5% (by anhydrous basis), crystals USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt
CAS Number:10034-99-8
:
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785
:246.48 g/mol
Merck Index:13,05715
Melting Pt:1124 °C
Density:1.67 g/cm³
Storage Temperature:Ambient
EINECS:231-298-2
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- GMP Manufactured Product:Passes Test
- Meets B.P. Chemical Specifications:5.0 - 9.2
- Meets E.P. Chemical Specifications:40.0 - 52.0%
- Meets U.S.P Requirements:≤0.014%
- Preserve in well-closed containers.:≤0.5 ppm
- CAUTION: For Manufacturing, processing or repackaging:≤0.003%
- Bulk Pharmaceutical Chemical:99.0 - 100.5%
- USP - Identification:Passes Test
- USP - pH (1 in 20):5.0 - 9.2
- USP - Loss on Ignition:40.0 - 52.0 %
- USP - Chloride (Cl):≤ 0.014 %
- USP - Iron (Fe):≤ 0.5 ppm
- USP - Selenium (Se):≤ 0.003 %
- USP - Assay (MgSO₄) (anhydrous basis):99.0 - 100.5 %
- USP - Endotoxin Concentration (EU/g):≤ 2.5
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Chloride (Cl):≤ 300 ppm
- EP/BP - Arsenic (As):≤ 2 ppm
- EP/BP - Iron (Fe):≤ 20 ppm
- EP/BP - Loss on Drying:48.0 - 52.0 %
- EP/BP - Assay (MgSO₄) (anhydrous basis):99.0 - 100.5 %
- Solution Test:Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_BAKR2504-01 spec_BAKR2504-05 spec_BAKR2504-07 spec_BAKR2504-08
Product specification
Frequently Bought Together