Magnesium chloride hexahydrate 99.0-102.0% ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7791-18-6
:
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781
:203.3 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:117 °C (–H₂O of crystall.)
Density:1.57 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-094-6
Specification Test Results
- Assay:99.0 - 102.0 %
- Water insoluble matter:Max. 0.005 %
- Barium (Ba):Max. 0.005 %
- Calcium (Ca):Max. 0.01 %
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 0.005 %
- Manganese (Mn):Max. 5 ppm
- Sodium (Na):Max. 0.005 %
- Strontium (Sr):Max. 0.005 %
- Heavy metals (as Pb):Max. 5 ppm
- Nitrate (NO₃):Max. 0.001 %
- Phosphate (PO₄):Max. 5 ppm
- Sulfate (SO₄):Max. 0.002 %
- Ammonium (NH₄):Max. 0.002 %
Specification Test Results
Frequently Bought Together