
Barium chloride dihydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:10326-27-9
:
Formula:BaCl₂·2H₂O
MDL Number:MFCD00149154
:244.27 g/mol
Merck Index:14,00971
Boiling Pt:1560 °C (1013 hPa)
Melting Pt:960 °C
Density:3.86 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:233-788-1
UN:1564
ADR:6.1,III
:GHS06
Frequently Bought Together