Order Entry
Belgium
ContactUsLinkComponent
Dipotassium hydrogen orthophosphate trihydrate, EMSURE® for analysis, Supelco®
Dipotassium hydrogen orthophosphate trihydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
Dipotassium hydrogen orthophosphate trihydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    340 °C decomposes,(anhydrous substance)
  • Molar Mass:
    228.23 g/mol
  • Bulk Density:
    800 kg/m3
  • pH:
    9.2 - 9.4 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together