Dipotassium hydrogen orthophosphate trihydrate, Reagent Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16788-57-1
:7778-77-0
Formula:K₂HPO₄·3H₂O
MDL Number:MFCD00149926
:228.22 g/mol
Merck Index:14,7658
Boiling Pt:∼340 °C (1013 hPa)
Density:2.504 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-834-5
Related Information
Frequently Bought Together