Order Entry
Belgium
ContactUsLinkComponent
Trisodium phosphate dodecahydrate ≥98% ACS, Fluka™
Trisodium phosphate dodecahydrate ≥98% ACS, Fluka™
Certificates
Trisodium phosphate dodecahydrate ≥98% ACS, Fluka™

Specification Test Results

  • Assay:
    98.0 - 102.0 %
  • Insoluble matter:
    Max. 0.01 %
  • Free alkali (as NaOH):
    Max. 2.5 %
  • Iron (Fe):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.01 %

Specification Test Results

Frequently Bought Together