
Trisodium phosphate dodecahydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:10101-89-0
:
Formula:Na₃PO₄·12H₂O
MDL Number:MFCD00149198
:380.12 g/mol
Merck Index:13,08736
Melting Pt:∼75 °C
Density:1.62 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-509-8
ADR:8,III
REACH:01-2119489800-32-xxxx
:GHS07
Frequently Bought Together